Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Pentahydrate, Powder, USP, 98.5-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7758-99-8
CuH10O9S
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
7758-99-8
CuH10O9S
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

CU110, 7758-99-8, CuSO4•5H2O

Spectrum™ Cupric Sulfate, Pentahydrate, Powder, USP is used as a mineral supplement and in topical antifungal preparations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
UN Number UN3077
Percent Purity 98.5 to 100.5%
Calcium (Ca) 5e-005
Loss on Drying 33.0-36.5%
Nickel (Ni) 5e-005
Packaging Poly Bottle
Potassium (K) 0.0001
Sodium (Na) 0.0002
Quantity 500 g