Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate Pentahydrate, Purified (Copper (II) Sulfate Pentahydrate), Reagents

Catalog No. 01337302
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg

Cupric Sulfate Pentahydrate, Purified (Copper (II) Sulfate Pentahydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym Copper(II) Sulfate Pentahydrate, Blue Vitriol
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Boiling Point >150°C
Melting Point 110°C
Color Blue
pH 4
Physical Form Solid
Quantity 50 kg
Grade Pure
Packaging Fiber Drum
Specific Gravity 2.286
Chemical Name or Material Cupric Sulfate Pentahydrate, Purified (Copper (II) Sulfate Pentahydrate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.