Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cuprous Chloride, Powder, ACS, 90%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7758-89-6
7758-89-6
Grade ACS
CAS Max % 1
Percent Purity 0.9
Calcium (Ca) 0.0001
Insoluble in Acid 0.0002
Iron (Fe) 5e-05
Packaging Amber Glass Bottle
Potassium (K) 0.0002
Sodium (Na) 0.0005
Sulfate 0.001
Quantity 125 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.