Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Curcumin, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
458-37-7
458-37-7
Packaging Amber Glass Bottle
Quantity 10 g
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.