Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Curcumin, Tocris Bioscience™

Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
458-37-7
C21H20O6
368.39
MFCD00008365
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
969516
CHEBI:3962
(1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
This item is not returnable. View return policy
458-37-7
C21H20O6
368.39
MFCD00008365
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
969516
CHEBI:3962
(1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O

Antitumor, anti-inflammatory and antioxidant

Antitumor, anti-inflammatory and antioxidant agent. Downregulates expression of reactive-oxygen-generating enzymes (cyclooxygenase, lipoxygenase, iNOS), TNFα, IL-1, IL-6, PKC, EGFR, NF-κB, IκB kinase and more. Upregulates expression of PPARγ, p53, Nrf2. Also displays antimicrobial, antidiabetic neuro- and cardioprotective properties in vivo.
Quantity 50 mg
Formula Weight 368.38
Percent Purity >98%
Chemical Name or Material Curcumin

Recommended Storage : Store at -20°C