Learn More
CurranTaylor™ Scotsman™ Benchtop Ice and Water Dispenser Products for Science Education
Use 40% less energy and 50% less water than standard cube ice machines! The CurranTaylor™ Scotsman™ Benchtop Ice and Water Dispenser is easy to clean and ideal for laboratory and clinical use. Sustainable and environmentally friendly design.
Manufacturer:  CurranTaylor™ BTD312A1/EMD
Catalog No. S28644
Add to cart
Description
Description
Makes nugget ice - dispenses easily and cools quickly
  • Durable stainless steel evaporator, greaseless bearings for reduced maintenance
  • Larger dispensing area to accommodate ice buckets, antimicrobial AquaArmor with AgION™ molded into key components
  • Sleek stainless steel finish – Easy to clean, ideal for laboratory and clinical use
  • Original nugget ice packs tightly and cools quickly
  • Makes up to 260 lb./day and stores 12 lb.
  • Removable front panels allow for easy access and service to all key internal components
  • Intuitive LED diagnostic lights reduce service time
  • Unit specific QR code for quick access to service manuals and warranty history
  • Convenient USB drive that allows for fast software updates
  • Quieter and hands free operation
  • Sustainable and environmentally friendly - Uses up to 40% less energy and 50% less water than standard cube ice machines
  • Outstanding factory warranty with exclusive CurranTaylor support
  • Cat. No. 09-540-121 filter package recommended
Specifications
Specifications
115V, 60Hz, 1 Phase, 15A
62cm
3 years (parts and labor on all components) and 5 years (parts on compressor and condenser)
89cm
Bench Nugget Ice Maker/Dispenser
24.38 in.
35 in.
41.2cm