Learn More
CurranTaylor™ Scotsman™ Filters and Filter Cartridges for Ice Makers Available on GSA/VA Contract for Federal Government customers only.
Scotsman™ Filters and Filter Cartridges for Ice Makers keeps ice makers cleaner while reducing failures due to lime scale, dirt, and excessive chlorine.
Manufacturer:  CurranTaylor™ HF20S
 View more versions of this product
Catalog No. 09-540-124
Add to cart
Description
Description
  • IMPACT media package in large cartridges
  • Built in scale inhibition
  • SQC™ (Sanitary Quick Change) cartridge
  • BSPT horizontal inlet and outlet port
  • 0.2 micron media
  • Valve-in-head
  • Optional pressure gauge
Benifits:
  • Higher flow rates, no need for manifold systems or for separate pre-filter and housing
  • Reduces ability of calcium and magnesium to precipitate on evaporator plates as hard scale
  • Allows fast and easy change-outs with 1/4 turn while minimising the possibility of leakage and contamination
  • Allows direct or easily adaptable connections to existing plumbing lines
  • Reduces microbial content in water and protects customers from cyst and 99.9% of common pathogens
  • Combined sediment reduction with chlorine, taste and odour reduction for high dirt loading applications
  • No separate shut off valve required, easy to change using un-skilled staff
  • Easy to monitor filter blockage for identification of time to change filter
  • Independent qualification of performance and guarantee of product safety
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
34069L (9000 gal.)
Reduces sediment and chlorine for equipment protection. Helps maintain efficiency with annual change.
Built-in scale inhibitor, bacteriostatic Carbon block filtration medium
34069L
NSF, FDA CFR-21
Ice makers
9000 gal.
Filter Cartridge for Model 121