Learn More
CurranTaylor™ Scotsman™ Filters and Filter Cartridges for Ice Makers
GSA_VASDP
Scotsman™ Filters and Filter Cartridges for Ice Makers keeps ice makers cleaner while reducing failures due to lime scale, dirt, and excessive chlorine.
$171.53 - $669.00
Specifications
Certifications/Compliance NSF, FDA CFR-21
For Use With (Equipment) Ice makers
View More Specs
Products 6
Catalog Number Mfr. No. Capacity For Use With (Application) Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Capacity For Use With (Application) Product Type Includes Price Quantity & Availability  
09-540-123
SDP
View Documents
CurranTaylor™
IMF160S
132489L (35000 gal.) Provides consistent high quality water for commercial ice by reducing the effects of sediment, chlorine, and odor and scale Large System Filter Package Built-in scale inhibitor, sanitary quick change encapsulated cartridge, 0.5 in. FNPT horizontal inlet and outlet ports, head assembly with integral mounting bracket, pressure gauge and single cartridge filter, NSF performance data sheet
Each for $442.27
Add to cart
09-540-125
SDP
View Documents
CurranTaylor™
RC40S
94635L (25000 gal.) Reduces sediment and chlorine for equipment. Helps maintain efficiency with annual change. Filter Cartridge for Model 122 Built-in scale inhibitor, bacteriostatic Carbon block filtration medium
Each for $171.53
Add to cart
09-540-121
GSA_VASDP
View Documents
CurranTaylor™
IMF120S
34069L (9000 gal.) Provides consistent high quality water for commercial ice by reducing the effects of sediment, chlorine, and odor and scale Small System Filter Package Built-in scale inhibitor, sanitary quick change encapsulated cartridge, 0.065 in. FNPT horizontal inlet and outlet ports, head assembly with integral mounting bracket, pressure gauge and single cartridge filter, internal automatic 0.25 turn shut-off valve, NSF performance data sheet
Each for $523.00
Edge
Add to cart
09-540-122
SDP
View Documents
CurranTaylor™
IMF140S
94635L (25000 gal.) Provides consistent high quality water for commercial ice by reducing the effects of sediment, chlorine, and odor and scale Medium System Filter Package Built-in scale inhibitor, sanitary quick change encapsulated cartridge, 0.065 in. FNPT horizontal inlet and outlet ports, head assembly with integral mounting bracket, pressure gauge and single cartridge filter, internal automatic 0.25 turn shut-off valve, NSF performance data sheet
Each for $669.00
Add to cart
09-540-124
GSA_VASDP
View Documents
CurranTaylor™
RC20S
34069L (9000 gal.) Reduces sediment and chlorine for equipment protection. Helps maintain efficiency with annual change. Filter Cartridge for Model 121 Built-in scale inhibitor, bacteriostatic Carbon block filtration medium
Each for $228.00
Add to cart
09-540-126
SDP
View Documents
CurranTaylor™
RC60S
132489L (35000 gal.) Reduces sediment and chlorine for equipment protection. Helps maintain efficiency with annual change. Filter Cartridge for Model 123 Built-in scale inhibitor, bacteriostatic Carbon block filtration medium
Each for $404.00
Add to cart
Description
Description
  • IMPACT media package in large cartridges
  • Built in scale inhibition
  • SQC™ (Sanitary Quick Change) cartridge
  • BSPT horizontal inlet and outlet port
  • 0.2 micron media
  • Valve-in-head
  • Optional pressure gauge
Benifits:
  • Higher flow rates, no need for manifold systems or for separate pre-filter and housing
  • Reduces ability of calcium and magnesium to precipitate on evaporator plates as hard scale
  • Allows fast and easy change-outs with 1/4 turn while minimising the possibility of leakage and contamination
  • Allows direct or easily adaptable connections to existing plumbing lines
  • Reduces microbial content in water and protects customers from cyst and 99.9% of common pathogens
  • Combined sediment reduction with chlorine, taste and odour reduction for high dirt loading applications
  • No separate shut off valve required, easy to change using un-skilled staff
  • Easy to monitor filter blockage for identification of time to change filter
  • Independent qualification of performance and guarantee of product safety
Specifications
Specifications
NSF, FDA CFR-21
Ice makers