Learn More
CurranTaylor™ Scotsman™ Prodigy™ Undercounter Cuber with 80 lbs. Storage Available on GSA/VA Contract for Federal Government customers only.
Scotsman™ Prodigy™ Undercounter Cuber uses energy efficiently, exceeding California and Federal energy efficiency regulations.
Manufacturer:  CurranTaylor™ CU1526SA
 View more versions of this product
Catalog No. 09-540-116
Add to cart
Alert:
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Description
Description
  • Cube ice maker machine
  • AutoAlert™ control panel communicates operating status and need for upkeep
  • The patented WaterSense adaptive purge control delivers maximum reliability by reducing scale buildup for a longer time between cleanings
  • Stainless steel external panels
  • Easily-removed door, top panel and storage bin allowing clear access to all internal components
  • A diagnostic code display insuring the right fix the first time and user-friendly front removable cleanable air filter are standard equipment
  • 80 lbs. of storage at a convenient level
  • Breathes in and out the front. No need to allow space at side or back for air circulation
  • Outstanding factory warranty with exclusive CurranTaylor support
  • Cat. No. 09-540-121 filter package recommended
09-540-116: 24 Hour Volume Production: 150 lbs. (or 110 lbs. ARI) air-cooled, 175 lbs. (or 124 lbs. ARI) water-cooled
09-540-117: 24 Hour Volume Production: 200 lbs. (or 155 lbs. ARI) air-cooled, 240 lbs. (or 190 lbs. ARI) water-cooled
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
15A
27.3 in.
115V 60Hz
33 in.
60Hz
Stainless Steel
115V
18 gal./100 lb.
68kg
60cm
80 lb.
Self Contained Cube Ice Maker
80 lb., 36kg
70cm
Uses energy efficiently, exceeding California and federal energy efficiency regulations
83.8cm
AutoAlert™ control panel, patented WaterSense™ adaptive purge control, Stainless Steel external panels, easily-removed door, top panel and storage bin, diagnostic code display
11.5kWh/100 lb.
3 Years
150 lbs.
26 in.
24 Hours Volume production 150 lb. (or 110 lb. ARI) air cooled, 175 lb. (or 124 lb. ARI) water cooled
36kg
Documents
Accessories