Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Current Technologies Dispensers for Bio-Screen™ or Fisherbrand™ BloodBloc™ Wipes
Reduce clutter on workbench, counters, trays, carts with either clear acrylic or white cardboard dispensers; sized specifically to store BloodBloc or Bioscreen wipes
Supplier: Current Technologies BH30000
This item is not returnable.
View return policy
Description
For 3 × 3 in. BloodBloc (06-670-1, -36, -49), and Bio-Screen (06-670, 14-412-52C, -52H) biohazard wipes
- Measures 3W x 3L x 3 in.H
- Measures 4W × 4L × 5.5 in.H
- May also be used with full-size HYPE-WIPE™ bleach towelettes, 14-412-56
Specifications
Cardboard Dispenser | |
3 x 4 x 4 in. (7.62 x 16.5 x 10.16 cm) | |
7.62 cm | |
16.5 cm | |
10.16 cm | |
Reduce clutter on workbench, counters, trays, carts |
White | |
3 in. | |
4 in. | |
4 in. | |
Cardboard | |
For 3 x 34 in. Bio-Screen or Fisherbrand BloodBloc wipes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction