Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Current Technologies Dispensers for Bio-Screen or Fisherbrand BloodBloc Wipes
Reduce clutter on workbench, counters, trays, carts with either clear acrylic or white cardboard dispensers; sized specifically to store BloodBloc or Bioscreen wipes
$7.80 - $26.75
Specifications
For Use With (Application) | Reduce clutter on workbench, counters, trays, carts |
---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W x H) | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W x H) | Material | Price | Quantity & Availability | |||
14-412-51
|
Current Technologies
ACBH33 |
Clear | 3.5 x 9.25 x 3.5 in. (8.89 x 2.54 x 8.89cm) | Acrylic |
Each for $26.75
|
|
14-412-51A
|
Current Technologies
BH10000 |
White | 4 x 10.25 x 4 in. (10.16 x 16.5 x 10.16cm) | Cardboard |
Pack of 10 for $7.80
|
|
14-412-51B
|
Current Technologies
BH30000 |
White | 3 x 4 x 4 in. (7.62 x 16.5 x 10.16cm) | Cardboard |
Pack of 10 for $7.80
|
|
Description
For 3 x 3 in. BloodBloc (06-670-1, -36, -49), and Bio-Screen (06-670, 14-412-52C, -52H) biohazard wipes
- Measures 3W x 3L x 3 in.H
- Measures 4W x 4L x 5.5 in.H
- May also be used with full-size HYPE-WIPE bleach towelettes, 14-412-56
Specifications
Reduce clutter on workbench, counters, trays, carts |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title