Learn More
HCL Labels Personalized Satellite Accumulation - 3" x 5" - Pack of 50
SDP
Supplier:  HCL Labels HWSA0003
Encompass_Preferred
This label is designed to help you comply with the labeling requirements for hazardous waste stored at or near the point of generation and defined as "satellite accumulation area" (SAA) see 40 CFR 262.34(c)(1). Send in your personalized information to be pre-printed.
Catalog No. NC1975517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.