Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety™ Cut Pro™ 92773

MCR Safety™ Cut Pro™ 92773 features Hypermax™ engineered yarn, which provides high levels of cut resistance.

$85.49

Specifications

Coating Material Polyurethane
Cut Resistance Rating ANSI A4
Color White
Material HPPE
For Use With (Application) Assembly production where extreme sense of touch is needed
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-812-769
View Documents
MCR Safety™
92773S
Small
Dozen of 12 for $85.49
Add to cart
 
19-812-770
View Documents
MCR Safety™
92773M
Medium
Dozen of 12 for $85.49
Add to cart
 
19-812-771
View Documents
MCR Safety™
92773L
Large
Dozen of 12 for $85.49
Add to cart
 
19-812-772
View Documents
MCR Safety™
92773XL
X-Large
Dozen of 12 for $85.49
Add to cart
 
19-812-773
View Documents
MCR Safety™
92773XXL
2X-Large
Dozen of 12 for $85.49
Add to cart
 
Description

Description

  • 15 gauge
  • White Hypermax™ engineered yarn shell
  • Excellent dexterity and flexibility
  • White PU palm and finger coating
  • Good abrasion resistance and grip
  • Comfortable and easy to wear
Specifications

Specifications

Polyurethane
White
Assembly production where extreme sense of touch is needed
Specific
12 pairs
ANSI A4
HPPE
Knitted
ANSI 3
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit