Learn More
Electron Microscopy Sciences Polypropylene Cutting Board
Supplier: Electron Microscopy Sciences 63308
This cutting board is autoclavable, unbreakable, non-absorbent, and easy to clean. It is a useful item in any laboratory. Our cutting boards are made from pure Polypropylene which can be autoclaved repeatedly. Polypropylene can withstand temperatures up to 135 Degrees F. Polypropylene can be sterilized. Made in the US of NSF, FDA and USDA approved material. Measurements: 300mm(L) x 210mm(W) x 12mm(H).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.