Learn More
Aviva Systems Biology Corp CUX2 Antibody - N-terminal region (ARP47629_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47629_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CUX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CUX2. Alternate names: CDP2, CUTL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59614