Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Steris™ CV™ Medicated Lotion Soap

Formulated to be effective against both gram-positive and gram-negative bacteria

Specifications

For Use With (Application) Effective against MRSA and VRE
View More Specs

Requires: 14-415-33 requires use of Steris Dispensing System (14-415-21).


Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
14-415-33
View Documents
Steris™
626287
1 L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
14-415-34
View Documents
Steris™
6262R2
444 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Contains 0.5% triclosan
  • Contains emollients and skin conditioners to be gentle on hands

Specifications

Specifications

Effective against MRSA and VRE
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit