Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CWC27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554184
Description
Immunogen sequence: REDQTLALLN QFKSKLTQAI AETPENDIPE TEVEDDEGWM SHVLQFEDKS RKVKDASMQD SDTFEIYDPR NPVNKRRREE SK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
As part of the spliceosome, plays a role in pre-mRNA splicing (PubMed:29360106). Probable inactive PPIase with no peptidyl-prolyl cis-trans isomerase activity (PubMed:20676357). [UniProt].
Specifications
CWC27 | |
Polyclonal | |
Unconjugated | |
Cwc27 | |
3110009E13Rik; Antigen NY-CO-10; CWC27; CWC27 spliceosome associated protein homolog; CWC27 spliceosome-associated protein homolog; CWC27 spliceosome-associated protein homolog (S. cerevisiae); NY-CO-10; peptidyl-prolyl cis-trans isomerase CWC27 homolog; peptidyl-prolyl cis-trans isomerase SDCCAG10; PPIase CWC27; PPIase SDCCAG10; Probable inactive peptidyl-prolyl cis-trans isomerase CWC27 homolog; SDCCAG10; SDCCAG-10; Serologically defined colon cancer antigen 10; Spliceosome-associated protein CWC27 homolog; UNQ438/PRO871 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10283, 361887, 67285 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3TKY6, Q5XIB2, Q6UX04 | |
Cwc27 | |
Recombinant protein corresponding to Human CWC27. Recombinant protein control fragment (Product #RP-91750). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction