Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CWC27 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549307

Catalog No. PA549307

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and monkey based on sequence homology.

PPIases accelerate the folding of proteins.
Specifications

Specifications

CWC27
Polyclonal
PBS with 0.09% sodium azide
Q6UX04
Peptidyl-prolyl cis-trans isomerase CWC27 homolog, PPIase CWC27, Antigen NY-CO-10, Serologically defined colon cancer antigen 10, CWC27, SDCCAG10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10283
Western Blot
Unconjugated
CWC27
Liquid
CWC27
KLH conjugated synthetic peptide between 411-437 amino acids from the C-terminal region of human CWC27.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit