Learn More
Medchemexpress LLC CWI1-2 | 2408590-36-1 | C22H17Cl3N6O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1532745MG
Encompass_Preferred
CWI1-2 is an IGF2BP2 inhibitor that binds IGF2BP2 and inhibits its interaction with m6A-modified target transcripts. It induces apoptosis and differentiation, and demonstrates promising anti-leukemic effects.
  • Binds to IGF2BP2
  • Inhibits interaction with m6A-modified target transcripts
  • Induces apoptosis and differentiation
  • Shows promising anti-leukemic effects
  • Exhibits antiproliferative activity with an IC50 of 200 nM against human THP-1 cells
  • Reduces glutamine uptake and impairs mitochondrial function in AML cells
  • Significantly inhibits the colony-forming ability of leukemic mouse blasts
  • Greatly impairs the self-renewal of leukemic stem/initiating cells
Catalog No. 50-003-72593