Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CX3CL1/Fractalkine Rabbit anti-Human, Clone: 22, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | CX3CL1/Fractalkine |
---|---|
Clone | 22 |
Dilution | Sandwich ELISA Capture 1:250-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Human samples. It is validated for Sandwich ELISA Capture.Specifications
CX3CL1/Fractalkine | |
Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
Recombinant Human CX3CL1/Fractalkine Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
22 | |
ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
0.2 um filtered solution in PBS | |
6376 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title