Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CXADR Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Specifications

Target CXADR
Conjugate HRP
Target Species Human
Gene ID (Entrez) 1525
Gene Symbol CXADR
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB009631
View Documents
Novus Biologicals™
NBP27929615PLATES
15 Plates N/A
NB009632
View Documents
Novus Biologicals™
NBP2792965PLATES
5 Plates N/A
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CXADR.
Specifications

Specifications

CXADR
Human
CXADR
HRP
1525
Rabbit
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit