Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXADR Rabbit anti-Human, Clone: 271, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | CXADR |
---|---|
Clone | 271 |
Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100, Sandwich ELISA Detection 1:1000-1:10000 |
Applications | Flow Cytometry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Description
CXADR Monoclonal specifically detects CXADR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection.Specifications
CXADR | |
Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100, Sandwich ELISA Detection 1:1000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3-binding protein, hCAR, HCVADR | |
Recombinant Human CXADR protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
271 | |
Flow Cytometry, Immunofluorescence, ELISA | |
Unconjugated | |
Rabbit | |
Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites | |
0.2 um filtered solution in PBS | |
1525 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title