Learn More
CXCL1/GRO alpha/KC/CINC-1 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0201-13H12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212216AF532
Catalog No. NP212216A32
Add to cart
Description
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal antibody specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CXCL1/GRO alpha/KC/CINC-1
RM0201-13H12
Western Blot
CXCL1
Mouse GRO alpha
0.1 mL
Primary
2919
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse