Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL1/GRO alpha/KC/CINC-1 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0201-13H12, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212216AF750

Catalog No. NP212216A70


Add to cart

Description

Description

CXCL1/GRO alpha/KC/CINC-1 Monoclonal antibody specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

CXCL1/GRO alpha/KC/CINC-1
RM0201-13H12
Western Blot
Conjugated, Purified
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CXCL1
Mouse GRO alpha
Protein G purified
Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Membrane Trafficking and Chaperones, Signal Transduction
Primary
2919.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit