Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL1 Mouse anti-Human, Clone: 2A10, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen CXCL1
Clone 2A10
Concentration 1 mg/mL
Applications Western Blot, Flow Cytometry
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC1705100
View Documents
MilliporeSigma
MABC1705100UG
100 μg
Each for $373.00
Add to cart
 
MABC170525
View Documents
MilliporeSigma
MABC170525UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

CXCL1 Monoclonal antibody specifically detects CXCL1 in Human samples. It is validated for Western Blotting, Flow Cytometry.
Specifications

Specifications

CXCL1
1 mg/mL
Unconjugated
Mouse
Human
Full-length mature recombinant human C-X-C motif chemokine 1 (CXCL1).
Monoclonal
Protein G purified
2A10
Western Blot, Flow Cytometry
Purified
RUO
Purified mouse monoclonal antibody IgG2b in PBS with 0.05% sodium azide.
IgG2bκ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit