Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL10 (IP10) Armenian Hamster anti-Mouse, Clone: 1B9, MilliporeSigma™

Armenian Hamster Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen CXCL10 (IP10)
Clone 1B9
Concentration 0.5 mg/mL
Applications ELISA, Inhibition Assays, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2342100
View Documents
MilliporeSigma
MABF2342100UL
100 μL
Each for $387.00
Add to cart
 
MABF234225
View Documents
MilliporeSigma
MABF234225UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

CXCL10 (IP10) Monoclonal antibody specifically detects CXCL10 (IP10) in Mouse samples. It is validated for ELISA, Inhibition, Western Blotting.
Specifications

Specifications

CXCL10 (IP10)
0.5 mg/mL
Unconjugated
Armenian Hamster
Mouse
Full-length mature recombinant murine CXCL10 (IP10) produced in E. coli.
Monoclonal
Protein G purified
1B9
ELISA, Inhibition Assays, Western Blot
Purified
RUO
Purified Amenian Hamster monoclonal antibody IgG1 in PBS without azide.
IgG1κ
Primary
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit