Learn More
Abnova™ Cxcl10 (Mouse) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Manufacturer:  Abnova™ KA5709
Catalog No. 89-339-640
Add to cart
Description
Description
Cxcl10 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of mouse Cxcl10.
Specifications
Specifications
Quantitative
Cxcl10
1 Kit
RUO
Mouse
15945
Standard curve
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
C7, CRG-2, INP10, IP-10, IP10, Ifi10, Scyb10, gIP-10, mob-1