Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CXCL11/I-TAC Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793115PLATES

 View more versions of this product

Catalog No. NB009662

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human CXCL11/I-TAC.
Specifications

Specifications

HRP
Antibody Pairs
12.5 pg/mL
6373
RUO
CXCL11/I-TAC
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research
Human
CXCL11
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 800 pg/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes an antibody specific for Human CXCL11/I-TAC.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human CXCL11/I-TAC polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of mouse anti-Human CXCL11/I-TAC monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 35 ng of recombinant Human CXCL11/I-TAC.
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    12.5-800 pg/mL
    Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit