Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CXCL11/I-TAC Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

$629.00

Specifications

Target CXCL11/I-TAC
Conjugate HRP
Target Species Human
Gene ID (Entrez) 6373
Gene Symbol CXCL11
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB009661
View Documents
Novus Biologicals™
NBP27931115PLATES
15 Plates
Each for $629.00
Add to cart
 
NB009662
View Documents
Novus Biologicals™
NBP2793115PLATES
5 Plates This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human CXCL11/I-TAC.
Specifications

Specifications

CXCL11/I-TAC
Human
CXCL11
HRP
6373
Rabbit/Mouse
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit