Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL11/I-TAC Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0203-17L2, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212220AF532

Catalog No. NP212220A32


Add to cart

Description

Description

CXCL11/I-TAC Monoclonal antibody specifically detects CXCL11/I-TAC in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

CXCL11/I-TAC
RM0203-17L2
Western Blot
Conjugated, Purified
CXCL11
Mouse CXCL11
Protein G purified
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research
Primary
6373.0
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit