Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
CXCL11/I-TAC Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0203-17L2, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212220AF750
Catalog No. NP212220A70
Description
Description
CXCL11/I-TAC Monoclonal antibody specifically detects CXCL11/I-TAC in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CXCL11/I-TAC
RM0203-17L2
Western Blot
Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CXCL11
Mouse CXCL11
0.1 mL
Primary
6373