Learn More
Aviva Systems Biology Corp CXCL12 Antibody - middle region (AVARP07031_P050) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP07031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CXCL12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CXCL12. Alternate names: IRH, PBSF, SDF1, TLSF, SDF1A, SDF1B, TPAR1, SCYB12ApplicationWB. Species reactivity: Cow, Dog, Pig, Rat
Catalog No. 50-014-62548