Learn More
CXCL12/SDF-1 Mouse anti-Human, Sheep, Alexa Fluor™ 532, Clone: MM0211-9N26, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212221AF532
Catalog No. NP212221A32
Add to cart
Description
Description
CXCL12/SDF-1 Monoclonal antibody specifically detects CXCL12/SDF-1 in Human,Sheep samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CXCL12/SDF-1
MM0211-9N26
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Sheep
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CXCL12
Human SDF1
0.1 mL
Primary
6387