Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL14/BRAK Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0213-12B24, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212224AF750

Catalog No. NP212224A70


Add to cart

Description

Description

CXCL14/BRAK Monoclonal antibody specifically detects CXCL14/BRAK in Human samples. It is validated for Western Blot.
Specifications

Specifications

CXCL14/BRAK
MM0213-12B24
Western Blot
Conjugated, Purified
CXCL14
Human CXCL14
Protein G purified
Biologically Active Proteins
Primary
9547.0
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
BMAC, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit