Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL15/Lungkine Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0207-16M7, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212416AF350

Catalog No. NP212416A30


Add to cart

Description

Description

CXCL15/Lungkine Monoclonal antibody specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

CXCL15/Lungkine
RM0207-16M7
Western Blot
Conjugated, Purified
CXCL15
Mouse
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
lungkine, Scyb15, weche
Rat
IgG2
0.1 mL
Primary
20309.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit