Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL15/Lungkine Antibody (RM0207-16M7), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212416AF594
Description
CXCL15/Lungkine Monoclonal specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot.Specifications
CXCL15/Lungkine | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CXCL15 | |
Mouse recombinant CXCL15/Lungkine | |
0.1 mL | |
20309.0 | |
Store at 4°C in the dark. |
Western Blot | |
RM0207-16M7 | |
Western Blot | |
lungkine, Scyb15, weche | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction