Learn More
CXCL16 Antibody (256213), mFluor Violet 500 SE, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB976MFV500
Description
CXCL16 Monoclonal antibody specifically detects CXCL16 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOFSpecifications
| CXCL16 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
| 58191 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Neutralization, CyTOF | |
| 256213 | |
| Western Blot, Flow Cytometry, Neutralization, CyTOF-ready | |
| chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
| E. coli-derived recombinant human CXCL16, Asn30-Pro118, Accession # Q9H2A7.4 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |