Learn More
CXCL16 Rat anti-Human, Alexa Fluor 647, Clone: 256213, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB976R100UG
Catalog No. FAB976R100
Add to cart
This item is not returnable. View return policy
Description
Description
CXCL16 Monoclonal antibody specifically detects CXCL16 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CXCL16
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_071342
CXCL16
E. coli-derived recombinant human CXCL16 Asn49-Pro137 Accession # NP_071342
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human CXCL16 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha ,12/SDF-1 beta , 13, recombinant mouse CXCL1, 2, 6, 9, 10, 12/SDF-1 alpha , 13, 16, recombinant rat CXCL1, 2, 3/CINC-2 alpha , 3/CINC-2 beta , recombinant canine CXCL8, or recombinant porcine CXCL8 is observed.
Flow Cytometry
256213
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCL16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Rat
IgG2a
RUO
Primary
58191
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)