Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL16 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0208-15Q2, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212226AF594

Catalog No. NP212226A94


Add to cart

Description

Description

CXCL16 Monoclonal antibody specifically detects CXCL16 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

CXCL16
RM0208-15Q2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
CXCL16
Mouse CXCL16
Protein G purified
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research
Primary
58191.0
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit