Learn More
CXCL16 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0208-15Q2, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212226AF750
Catalog No. NP212226A70
Add to cart
Description
Description
CXCL16 Monoclonal antibody specifically detects CXCL16 in Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CXCL16
RM0208-15Q2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CXCL16
Mouse CXCL16
0.1 mL
Primary
58191