Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | CXCL17 |
---|---|
Concentration | 0.55 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5122057
|
Invitrogen™
PA5122057 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5122056
|
Invitrogen™
PA5122056 |
100 μL |
Each for $511.50
|
|
|||||
Description
CXCL17 is a chemokine that acts as chemoattractant for monocytes, macrophages and dendritic cells. In particular, CXCL17 is a mucosal chemokine that attracts dendritic cells and blood monocytes to the lungs. CXCL17 plays a role in angiogenesis and possibly in the development of tumors. Further, CXCL17 acts as an anti-inflammatory in the stomach and may play a role in the innate defense against infections. CXCL17 also activates the C-X-C chemokine receptor GPR35 to induce a rapid and transient rise in the level of intracellular calcium ions. 4-Cys CXCL17 seems to exhibit much higher chemoattractant potency on monocytes and macrophages than 6-Cys CXCL17. Diseases associated with CXCL17 dysfunction include cholera.Specifications
CXCL17 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6UXB2 | |
284340 | |
Leu24-Leu119 (Accession Q6UXB2), with N-terminal His and GST Tag. | |
Primary | |
-20°C | |
CXCL17 |
0.55 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
13.6 kDa protein; 13.6-kDa protein; 4-Cys CXCL17; 6-Cys CXCL17; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 17; CXC; CXC motif chemokine; C-X-C motif chemokine 17; C-X-C motif chemokine ligand 17; CXCL; CXCL17; Dcip1; Dendritic cell and monocyte chemokine-like protein; DMC; UNQ473; UNQ473/PRO842; Vcc1; VCC-1; VEGF coregulated chemokine 1; VEGF co-regulated chemokine 1 | |
CXCL17 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title