Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CXCL2/GRO beta/MIP-2/CINC-3 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2794915PLATES

 View more versions of this product

Catalog No. NB010022


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Mouse CXCL2/GRO beta/MIP-2/CINC-3.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
3.91 pg/mL
2920
RUO
CXCL2/GRO beta/MIP-2/CINC-3
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
Mouse
CXCL2
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 250 pg/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes an antibody specific for Mouse CXCL2/GRO beta/MIP-2/CINC-3.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-CXCL2/GRO beta/MIP-2/CINC-3 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Mouse CXCL2/GRO beta/MIP-2/CINC-3 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 7.3 ng of recombinant Mouse CXCL2/GRO beta/MIP-2/CINC-3.
  • Storage is content dependent.
    Rabbit
    5 Plates
    3.91-250 pg/mL
    chemokine (C-X-C motif) ligand 2, CINC-2a, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2a, MIP2A, MIP2-alpha, SCYB2
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit