Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL2/GRO beta/MIP-2/CINC-3 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen CXCL2/GRO beta/MIP-2/CINC-3
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093584
View Documents
Novus Biologicals
NBP303233100UL
100 μg Each for $320.71
Add to cart
 
NB093585
View Documents
Novus Biologicals
NBP30323320UL
20 μg Each for $138.37
Add to cart
 
Description

Description

CXCL2/GRO beta/MIP-2/CINC-3 Polyclonal antibody specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

CXCL2/GRO beta/MIP-2/CINC-3
Unconjugated
Rabbit
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
PBS with 50% glycerol, pH7.3.
2920
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, Western Blot
Purified
RUO
Human, Rat
chemokine (C-X-C motif) ligand 2, CINC-2a, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2a, MIP2A, MIP2-alpha, SCYB2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-107 of human CXCL2/GRO beta/MIP-2/CINC-3 (NP_002080.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit