Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL4/PF4 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0215-11N29, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212229AF350

Catalog No. NP212229A30


Add to cart

Description

Description

CXCL4/PF4 Monoclonal antibody specifically detects CXCL4/PF4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CXCL4/PF4
MM0215-11N29
Western Blot
Conjugated, Purified
PF4
Human PF4/Platelet factor 4
Protein G purified
Biologically Active Proteins, Cancer, Cardiovascular Biology, Signal Transduction
Primary
5196.0
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, platelet factor 4, SCYB4oncostatin-A
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit