Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL4/PF4 Antibody (RM0210-14M15), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212230AF594
Description
CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CXCL4/PF4 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PF4 | |
Mouse recombinant CXCL4/PF4 | |
0.1 mL | |
5196 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0210-14M15 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, platelet factor 4, SCYB4oncostatin-A | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction