Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL4/PF4 Rat anti-Mouse, Janelia Fluor 646, Clone: RM0210-14M15, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212230JF646
Description
CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CXCL4/PF4 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PF4 | |
Mouse recombinant CXCL4/PF4 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0210-14M15 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, platelet factor 4, SCYB4oncostatin-A | |
Rat | |
Protein A or G purified | |
Biologically Active Proteins, Cancer, Cardiovascular Biology, Signal Transduction | |
5196 | |
Store at 4°C in the dark. | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title