Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL5/ENA-78 Mouse anti-Human, Clone: MM0216-10L19, Novus Biologicals™
Mouse Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | CXCL5 |
---|---|
Clone | MM0216-10L19 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Blocking Assay |
Classification | Monoclonal |
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
CXCL5 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P42830 | |
6374 | |
IgG1 | |
Protein G purified |
MM0216-10L19 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Blocking Assay | |
Unconjugated | |
Mouse | |
Human | |
chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5 | |
CXCL5 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title