Learn More
CXCL5/ENA-78 Rat anti-Mouse, Alexa Fluor™ 350, Clone: 32B3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222026AF350
Catalog No. NP222026A30
Add to cart
Description
Description
CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
CXCL5/ENA-78
32B3
Western Blot, ELISA
chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Rat
IgG2b
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CXCL5
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse PlGF recombinant protein. Rat monoclonal anti Mouse CXCL5 P50228.
0.1 mL
Primary
6374