Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL5/ENA-78 Rat anti-Mouse, Alexa Fluor™ 594, Clone: 32B3, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222026AF594

Catalog No. NP222026A94


Add to cart

Description

Description

CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CXCL5/ENA-78
32B3
Western Blot, ELISA
Conjugated, Purified
CXCL5
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse PlGF recombinant protein. Rat monoclonal anti Mouse CXCL5 P50228.
Protein G purified
Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primary
6374.0
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Rat
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit