Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL5/ENA-78 Rat anti-Mouse, Clone: 32B3, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP2220260.025MG
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA, Block/Neutralize.Specifications
CXCL5 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:1000, ELISA 1:1000-1:2000, Blocking/Neutralizing 50-200 molar excess | |
P50228 | |
CXCL5 | |
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse PlGF recombinant protein. Rat monoclonal anti Mouse CXCL5 P50228. | |
0.025 mg | |
Cell Cycle and Replication | |
6374 | |
Mouse | |
Purified |
ELISA, Blocking Assay, Western Blot | |
32B3 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only